missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum powder, -40 mesh, 99.7% (REO)

Product Code. p-4824769
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15491967

Brand: Thermo Scientific Alfa Aesar 044182.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Air and moisture | CAS: 7439-91-0 | La | 138.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-91-0
Molecular Formula La
Molecular Weight (g/mol) 138.91
MDL Number MFCD00011066
InChI Key FZLIPJUXYLNCLC-UHFFFAOYSA-N
Synonym in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
PubChem CID 23926
ChEBI CHEBI:33336
IUPAC Name lanthanum
SMILES [La]

Specifications

Color Gray to Silver
Appearance Gray to silver powder
Quantity 5 g
Mesh Size -40 Mesh
Odor Odorless
Sensitivity Air and moisture sensitive
Assay Percent Range 99.9% (REO)
Form Powder
Chemical Name or Material Lanthanum

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.