missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals

Product Code. 11353828
Click to view available options
Quantity:
10 mm
25 mm
50 mm
100 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-91-0
Molecular Formula La
Molecular Weight (g/mol) 138.91
MDL Number MFCD00011066
InChI Key FZLIPJUXYLNCLC-UHFFFAOYSA-N
Synonym in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
PubChem CID 23926
ChEBI CHEBI:33336
IUPAC Name lanthanum
SMILES [La]

Specifications

Color Silver to Gray
UN Number UN3208
Quantity 25 mm
Percent Purity 99.9%
Packaging Mylar under Argon
Weight ≈0.80g/mm
Form Cast Rod
Assay (metals basis excluding Ta)
Chemical Name or Material Lanthanum rod, 12.7mm (0.5 in.) dia.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.