Learn More
Thermo Scientific™ LanthaScreen™ Eu-PY20 Antibody Kit
LanthaScreen™ Eu-PY20 Antibody Kit
Brand:  Thermo Scientific™ PV5696
 View more versions of this product
Product Code. 10003624
  • 10720.00 € / Each
Explore more special offers
Description
Description
  • A general reagent to monitor tyrosine kinase activity
  • The PY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate
  • For use in combination with AlexaFluor 647-poly-GT (PV5693) to evaluate inhibitors in medium to high throughput screening applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Europium/Lanthanide Chelates
Europium-labeled anti-PY20 antibody in HEPES buffered saline. Store at -20°C.
340/615
TR-FRET
LanthaScreen™
1 mg
Fluorescence
Dry Ice
EGFR