Learn More
Thermo Scientific™ LanthaScreen™ Tb-DUB Substrate
Deubiquitinating enzymes (DUBs) play housekeeping functions by maintaining pools of active ubiquitin, and regulatory function by rescuing specific target proteins from degradation by the proteasome.
Marca: Thermo Scientific™ PV4444
Descrizione
Deubiquitinating enzymes (DUBs) play housekeeping functions by maintaining pools of active ubiquitin, and regulatory function by rescuing specific target proteins from degradation by the proteasome. The LanthaScreen™ Tb-DUB Substrate is a general substrate for assaying DUB activity in a high throughput assay format.

Specifica
100μg | |
Fluorescence | |
Pharma and biopharma, target and lead identification and validation, drug discovery and development, proteasome biology, biochemical proteasome assays, epigenetics assays for drug discovery | |
100 μg | |
Protein-Based Substrate | |
340/490, 520 nm | |
LanthaScreen™ Tb-DUB Substrate, 100μg, Play Housekeeping Functions by Maintaining Pools of Active Ubiquitin and Regulatory Function by Rescuing Specific Target Proteins from Degradation by the Proteasome, It is a General Substrate For Assaying DUB | |
42.8kDa | |
Kinetic | |
–80°C |
LanthaScreen™ DUB substate in HEPES buffered saline. Store at -80°C. | |
Frozen | |
LanthaScreen™ | |
DUB | |
Deconjugating Enzyme Substrate | |
Microplate Reader | |
Tb (Terbium), GFP (EmGFP) | |
LanthaScreen™ Tb-DUB Substrate | |
Dry Ice | |
TR-FRET |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only. Not for use in diagnostic procedures.