missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-labeled Anti-Goat Ab
This product is ideally suited for use during assay development when evaluating multiple primary goat antibodies, or when it is not possible to directly terbium label a primary antibody.
872.00 € - 12090.00 €
Specifications
Content And Storage | Terbium-labeled anti-Goat antibody in HEPES buffered saline. Store at -20°C. |
---|---|
Conjugate | Lanthanide Chelates, Terbium (Tb) |
Product Line | LanthaScreen™ |
Excitation/Emission | 340/490, 520 |
Reactivity | Goat |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10666255
|
Thermo Scientific™
PV3772 |
1 mg |
12090.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10369244
|
Thermo Scientific™
PV3770 |
25 μg |
872.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- A general reagent for use in assay development, this terbium labeled antibody will bind goat antibodies
- Ideally suited for use during assay development when evaluating multiple primary goat antibodies
- Also for use when it is not possible to to directly terbium label a primary antibody
Specifications
Terbium-labeled anti-Goat antibody in HEPES buffered saline. Store at -20°C. | |
LanthaScreen™ | |
Goat | |
Dry Ice |
Lanthanide Chelates, Terbium (Tb) | |
340/490, 520 | |
End Point |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title