missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ Tb-labeled Anti-Mouse Antibody Kit

Product Code. p-7181445
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
1mg
25µg
This item is not returnable. View return policy

Product Code. 11593517

Brand: Invitrogen™ PV3765

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

To facilitate in-house assay development around multiple target classes and assay formats, Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents.

To facilitate in-house assay development around multiple target classes and assay formats, Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents. Tb-labeled secondary antibodies bind to their corresponding species IgG with low nanomolar apparent Kd's, and show negligible cross reactivity with IgG's from other species. Each Tb-labeled anti-species antibody is provided with a corresponding fluorescein-labeled IgG as a positive control.

In a kinase assay utilizing the Tb-labeled antibody generic reagent, anti-phosphopeptide primary antibody is not labeled and the detection step is performed by the addition of the Tb-labeled secondary antibody. This approach can be applied to other target classes and assay formats.

TRUSTED_SUSTAINABILITY

Specifications

Description LanthaScreen™ Tb-labeled Anti-Mouse Secondary Antibody Kit, 25μg, Liquid, Mouse Species Reactivity, Mouse Host, IgG Detection Target, Terbium Conjugate, Fluorescent Detection Method, Water Soluble, 6 Months Shelf Life, Store in Freezer, Used for
Content And Storage Store in freezer (-5°C to -30°C).
Detection Method Fluorescent
Quantity 25 μg
Reactivity Mouse
Shipping Condition Dry Ice
Conjugate Lanthanide Chelates, Terbium (Tb)
Target IgG
Format Liquid
Formulation Liquid
Kit Contents TR-FRET dilution buffer, LanthaScreen™ Tb-anti-mouse antibody, fluorescein-mouse IgG
For Use With (Application) Pharma and biopharma, secondary detection, target and lead identification and validation
Product Type LanthaScreen™ Tb-labeled Anti-Mouse Antibody Kit
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.