missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ Tb-labeled Anti-rabbit Ab

A general reagent for use in assay development, this terbium labeled antibody will bind rabbit antibodies.

Brand:  Invitrogen™ PV3774

 View more versions of this product

Product Code. 10359194

  • 848.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

A general reagent for use in assay development, this terbium labeled antibody will bind rabbit antibodies. This product is ideally suited for use during assay development when evaluating multiple primary rabbit antibodies or when you are unable to directly terbium label your primary antibody.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

LanthaScreen™ Tb-labeled Anti-Rabbit Secondary Antibody, 25μg, Liquid, 1.03mg/mL Concentration, Rabbit Species Reactivity, Terbium Conjugate, Fluorescent Detection Method, 6 Months Shelf Life, Store in –20°C, Used for Pharma and Biophar
Fluorescence
25 μg
End Point
Lanthanide Chelates, Terbium (Tb)
Liquid
LanthaScreen™ Tb-labeled Anti-rabbit Antibody
Terbium-labeled anti-Rabbit antibody in HEPES buffered saline.

Store at -20°C.
340/490, 520 nm
Rabbit
Dry Ice
Liquid
Pharma and biopharma, secondary detection, cell analysis, drug discovery and development, target and lead identification and validation, kinase biology, kinase antibodies and immunoassays, biochemical kinase assays, secondary detection
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.