Learn More
Thermo Scientific™ LanthaScreen™ Tb-PY20 Antibody Kit
A general reagent to monitor tyrosine kinase activity, the pY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate.
Brand:  Thermo Scientific™ PV3553
 View more versions of this product
Product Code. 10042672
  • 13690.00 € / 1mg
Explore more special offers
Description
Description
A general reagent to monitor tyrosine kinase activity, the pY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate.
For use in combination with Fluorescein-poly-GT (Cat. No. PV3610) or Fluorescein-poly-GAT (PV3611) to evaluate inhibitors in medium to high throughput screening applications.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tyrosine, phospho-20
Tb (Terbium)/FITC
1 mg
Protein Biology
1 Kit(s)
150,000 Da
340/490, 520 nm
End Point
Tyrosine Kinases
ABL1, ABL2, AXL, BLK, BMX, BTK, CSF1R, CSK, DDR2, EGFR, EPHA1, EPHA2, EPHA3, EPHA4, EPHA5, EPHA7, EPHA8, EPHB1, EPHB2, EPHB3, EPHB4, ERBB2, ERBB4, FER, FGFR1, FGFR2, FGFR3, FGFR4, FGR, FLT1, FLT3, FLT4, FRK, FYN, HCK, IGF1R, INSR, KDR, KIT, LCK, LTK, LYNA, LYNB, MATK, MERTK, MET, MST1R, MUSK, NTRK1, NTRK2, NTRK3, PDGFRA, PDGFRB, PTK2, PTK2B, PTK6, RET, ROS1, SRC, SRCN1, SRMS, SYK, TEC, TEK, TNK2, TTK, TYRO3, YES1, ZAP70
0.54mg/mL
150kDa
RUO
Primary
Terbium-labeled anti-pY20 antibody in HEPES buffered saline. Store at -20°C.
384-well Plate
Dry Ice
Signal Transduction
TR-FRET
LanthaScreen™