Learn More
Invitrogen™ LanthaScreen™ TR-FRET BACE1 Assay Kit
BACE1 (beta-secretase) is a key enzyme involved in the production of amyloid beta-peptides found in extracellular amyloid plaques of Alzheimer's disease (AD).
Brand: Invitrogen™ PV4748
Description
BACE1 (beta-secretase) is a key enzyme involved in the production of amyloid beta-peptides found in extracellular amyloid plaques of Alzheimer's disease (AD). In some cases, early-onset familial AD can be attributed to a 'Swedish' mutation in the amyloid precursor protein (APP), which dramatically enhances the cleavage of this protein by BACE1. This and other genetic and pathological evidence has led to therapeutic approaches that focus on the inhibition of BACE1 and other APP-cleaving enzymes, such as gamma-secretase.
Invitrogen's LanthaScreen™ TR-FRET BACE1 assay provides sensitive high-throughput screening for potential inhibitors of beta-secretase. The kit uses a terbium (Tb)-labeled anti-biotin antibody and a fluorescein-labeled BACE1-biotin substrate in a homogeneous TR-FRET assay format (Figure 1).
Contents and Storage:
The LanthaScreen™ TR-FRET BACE1 Assay Kit contains BACE1 protein, fluorescently labeled BACE1 substrate, Tb-labeled anti-biotin antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C).

Specifications
Store in freezer (-5°C to -30°C). | |
β-Secretase Assay | |
LanthaScreen™ TR-FRET BACE1 Assay Kit | |
Beta Secretase Substrate | |
TR-FRET | |
For research use only. Not for use in diagnostic procedures | |
-5°C to -30°C |
LanthaScreen™ TR-FRET BACE1 Assay Kit, 800 x 20μL, Liquid, β-Secretase Assay, Used in Protein Assays and Analysis, Peptide-Based Substrate Properties, Beta Secretase (BACE1) Target Enzyme, TR-FRET Technique, Beta Secretase Substrate | |
LanthaScreen™ | |
Peptide-Based Substrate | |
800 x 20μL | |
800 x 20 μL Assays | |
Dry Ice | |
Beta Secretase (BACE1) |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2587156 | Certificate of Analysis | 12/05/2023 | PV4748 |
2587156 (2) | Certificate of Analysis | 04/05/2023 | PV4748 |
25871546 | Certificate of Analysis | 04/05/2023 | PV4748 |
2124089 | Certificate of Analysis | 21/11/2019 | PV4748 |
1688172 | Certificate of Analysis | 16/04/2017 | PV4748 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.