Learn More
Invitrogen™ LanthaScreen™ TR-FRET RAR alpha Coactivator Assay Kit, goat
LanthaScreen™ TR-FRET RAR alpha Coactivator Assay Kit, goat
Brand:  Invitrogen™ PV4409
Product Code. 10302223
  • 1796.00 € / Pack of 800
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15122:
The LanthaScreen™ TR-FRET Retinoic Acid Receptor (RAR) alpha Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential RAR alpha ligands as agonists of coactivator recruitment. The homogeneous mix-and-read assay format results in a ligand EC50 composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and recruit coactivator peptide (Figure 1).
How it works
The LanthaScreen™ TR-FRET RAR alpha Coactivator Assay Kit includes a terbium (Tb)-labeled anti-GST antibody, a fluorescein-labeled coactivator peptide, and a human RAR alpha ligand-binding domain (RAR alpha-LBD) that is tagged with glutathione-S-transferase (GST). To assay, RAR alpha-LBD is added to ligand test compounds, followed by addition of a mixture of the fluorescein-coactivator peptide and Tb-anti-GST antibody. After room temperature incubation, the TR-FRET 520:495 emission ratio is calculated and used to determine the EC50 from a dose response curve of the compound.
Contents and Storage:
The LanthaScreen™ TR-FRET RAR alpha Coactivator Assay Kit contains RAR alpha-LBD (GST) protein, fluorescein-labeled D22 coactivator peptide, Tb-anti-GST antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tb (Terbium)
Microplate Reader
Store in ultra-cold freezer (-68 to -85°C).
5914
Dry Ice
LanthaScreen™
RAR alpha
800 x 20 μL assays
Fluorescence
384-well Plate
Biochemical coregulator interaction
End Point
RARA, RAR alpha, NR1B1
Co-Factor Interaction Assay, TR-FRET
TR-FRET RAR Alpha Coactivator Assay Kit