Learn More
OHAUS™ Large Three Prong Sleeves
OHAUS™ Large Three Prong Sleeves are for use with clamps and can be easily removed for cleaning or replacement.
Brand: OHAUS™ 30400265
This item is not returnable.
View return policy
Description
- Available in red or black vinyl and fiberglass
- Fiberglass sleeves are ideal for application above 100°C
Specifications
3 | |
Vinyl | |
Black |
Prong Sleeve | |
Large |