Learn More
Brady™ LASERTAB™ Laser Printable Labels, B-747 Polyester
Designed for use with laser printers. Brady LASERTAB Laser Printable Labels are made of B-747 polyester.
Brand:  Brady™ 029794
 View more versions of this product
Product Code. 11503644
  • 464.00 € / 1 pack
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Blank, printable polyester labels for Laser Printers
  • CSA Accepted material when printed with designated laser printer toners
Specifications
Specifications
Chemical, high temperature, humidity, smear, UV and weather-Resistant
Yellow
Polyester (B-747)
2500/Pk.
1.875 x 0.833 in. (W x H)
0.0037 in.
Acrylic, Permanent
Matte
Laser Printable Label
Rectangle
47.6 x 21.1 mm (W x H)