Learn More
Brady™ LASERTAB™ Laser Printable Labels, B-747 Polyester
Designed for use with laser printers. Brady LASERTAB Laser Printable Labels are made of B-747 polyester.
Brand:  Brady™ 029789
 View more versions of this product
Product Code. 11553634
  • 733.00 € / 1 pack
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Blank, printable polyester labels for Laser Printers
  • CSA Accepted material when printed with designated laser printer toners
Specifications
Specifications
LAT-7-747-10-YL
Acrylic, Permanent
Matte
Laser Printable Label
Rectangle
25.4 x 12.7 mm (W x H)
Chemical, high temperature, humidity, smear, UV and weather-Resistant
White
Polyester (B-747)
10,000/Pk.
1 x 0.5 in. (W x H)
0.0037 in.