Learn More
Brady™ LASERTAB™ Laser Printable Labels, B-747 Polyester
Designed for use with laser printers. Brady LASERTAB Laser Printable Labels are made of B-747 polyester.
Brand:  Brady™ 029700
 View more versions of this product
Product Code. 11583564
  • 295.00 € / 1 pack
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Blank, printable polyester labels for Laser Printers
  • CSA Accepted material when printed with designated laser printer toners
Specifications
Specifications
UL Recognised; CSA Accepted
Acrylic, Permanent
Matte
Laser
17.78 mm
10,000/Pk.
0.3 x 0.25 in. (W x H)
-40°C to +130°C
0.094 mm
7.62 mm
6.35 mm
Chemical, high temperature, humidity, smear, UV and weather-Resistant
White
Polyester (B-747)
Laser Printable Label
22
Rectangle
7.62 x 6.35 mm (W x H)
0.0037 in.
6.35 x 7.62 mm
6.35 mm