Learn More
Brady™ LaserTab Laser Printable Polyolefin Labels
Blank, printable labels suitable for use with Laser Printers.
Brand:  Brady™ 029760
 View more versions of this product
Product Code. 11513614
  • 453.00 € / 1 pack
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
LaserTab Labels are available in the following formats:
  • Tamper-evident polyolefin
  • Polyester (UL Recognized Component when printed with designated laser printer toners)
  • Self-laminating
  • Nylon cloth
  • Paper
  • Polyimide
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
Matte
Laser
Asset Tracking, Circuit Board Labeling, General Identification
25.4 mm
304
Rectangle
0.23 mm
20.3 mm
6.4 mm
LAT-4-410-10
White
Polyolefin (Tamper-evident)
Non-sterile
Laser Printers
8
10,234 Labels
-70°C to +100°C
20.3 mm
6.4 mm