Learn More
Novacyt - Microgen Bioproducts Latex Agglutination Test: Salmonella
Can be used to obtain rapid, presumptive-positive results from selective media for targeted identification/diagnosis.
Brand:  Novacyt - Microgen Bioproducts M42CE
Product Code. 15978012
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • From a range of latex products for the rapid diagnosis of the most common pathogens for foodborne illness.
  • Specific tests available for a variety of infection detections.
  • Rapid (2 minute), simple, sensitive, and specific latex agglutination test for the identification and confirmation of infecting colonies.
  • Can be used to obtain rapid, presumptive-positive results and provide an early indication of possible production process failure.
  • Can be used to screen colonies before full biochemical identification and can replace traditional agglutinating antibody methods.
Specifications
Specifications
2°C to 8°C
Latex Agglutination Test
2 min.
Salmonella
50 Tests