Learn More
Fisherbrand™ Latex Dropper Teats
For easy dispensing of liquids.
Brand:  Fisherbrand™ DT/1/DO01
 View more versions of this product
Product Code. 12416180
  • 44.00 € / Pack of 20
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Flexible rubber bulbs create suction and dispensing capacity when used with pipettes
  • Provide intuitive manual pipeting control
  • Available in different capacities
Specifications
Specifications
Latex
Bulb Dropper
1.5 mL
5 mm