missing translation for 'onlineSavingsMsg'
Learn More

Semperguard™ Latex Powder-Free Innercoated Gloves

Product Code. 15583506
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Semperguard™ Latex Powder-free Innercoated Gloves are disposable protective glove made of natural rubber with a synthetic inner coating.

  • CE category III protective glove for complex risks
  • CE Class I examination glove
  • Skin-friendly, powder-free hygiene glove made of natural rubber
  • Good grip and easy to don through innovative production technology
  • Free from phthalates/softeners
  • Tested and certified according to 1935/2004 - suitable for food contact

Specifications

Certifications/Compliance EN 420, EN 374, EN 455; Medical device class I; Single-use protective glove for complex risks, category III
Coated Area Inside
Coating Material Synthetic
Color Natural
Disposable Single-use
Exterior Finish Fully Textured
Hand Ambidextrous
Material Rubber
Powdered Powder Free
Product Type Protective Gloves
Size X-Small
Sterility Non-sterile
Thickness (Metric) 0.24 mm (measure in double layer)
For Use With (Application) Medical device class I. Suitable for food contact
Glove Cuff Style Rolled
Length (Metric) 240 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'