missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ LB Broth (1X)
GREENER_CHOICE

LB broth (Lennox formulation) is a ready-to-use liquid growth medium used for bacterial culture.

Brand:  Gibco™ 10855001

44.76 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 11380872

  • 52.25 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

LB broth (Lennox formulation) is a ready-to-use liquid growth medium used for bacterial culture. Gibco LB Broth is filter-sterilized and can be used for nearly all applications that require a liquid bacteria culturing step, such as protein expression, cloning, and expansion.

LB broth is one of the most referenced bacterial media for cultivating and maintaining E. coli strains. Our LB broth is based on the Lennox formulation, one of the most commonly used formulations for LB medium.

LB broth formulation

Formulation per liter: 10 g SELECT Peptone 140, 5 g SELECT Yeast Extract, 5 g sodium chloride.

  • Tryptone provides nitrogen, vitamins, minerals, and amino acids essential for growth
  • Yeast extract is a source of vitamins, particularly B-group vitamins
  • Sodium chloride supplies essential electrolytes for transport and osmotic balance

Primary applications

  • Growth and maintenance of E. coli strains for plasmid purification and/or protein expression
  • Resuspension of T. pyriformis cell fractions

Gibco media bottles

Each media bottle features an angled opening for simplified pipetting. The 1-L Gibco media bottle uses 39% less source material (33% less for the 500-mL bottle) than comparable media bottles on the market. The cap and bottle are made from high-density polyethylene (HDPE) and polyethylene terephthalate (PETE) and can both be recycled.

Find the media format that you need

We offer other ready-to-use media formulations, such as Terrific Broth (Cat. No. A1374301) or S.O.C Recovery Medium (Cat. No. 15544034).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Escherichia coli
Bottle
Ready-to-Use
Sterile
1 L
• 1 L LB Broth (1X)

Store at room temperature.

LB Broth (Lennox)
Liquid Medium
Coliforms
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ LB Broth (1X) > Quantity: 1 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.