missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals LB Broth (Miller's modification)

(+)-Dehydroisoandrosterone, CAS # 53-43-0, also known as androstenolone, is an endogenous steroid hormone precursor. It is an intermediate in the conversion of cholesterol to androgens and estrogens.

25.93 € - 64.12 €

Specifications

Form Powder
Sensitivity Hygroscopic
View More Specs

Products 2
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
11362599
View Documents
Thermo Scientific Chemicals
H26676.22
100 g Glass Bottle
25.93 €
100g
In Stock
to see stock.
Add to basket
 
11372599
View Documents
Thermo Scientific Chemicals
H26676.36
500 g Plastic Bottle
64.12 €
500g
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

For the growth of E. coli and other enteric bacteria. LB Luria contains less salt than LB Miller and LB Lennox. LB Luria is used for cultivating salt-sensitive strains of E. coli and enteric bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• (+)-Dehydroisoandrosterone is an endogenous steroid hormone precursor that acts as an intermediate in the conversion of cholesterol to androgens and estrogens
Applications
• It is an endogenous and naturally occurring steroid hormone produced largely by the adrenal glands, but also by the testes and ovaries
• It has been described that (+)-Dehydroisoandrosterone could have a role in the immune and stress response
• (+)-Dehydroisoandrosterone can be converted to testosterone, androstenedione, estradiol, and estrone
• (+)-Dehydroisoandrosterone is derived from cholesterol through the action of two cytochrome P450 enzymes
• It can induce both direct and indirect in vivo biological effects, by modulating GABA- α receptor and N-Methyl-D-aspartic acid (NMDA) receptor complexes.
• It can also increase pancreatic β cell insulin secretion and anti-glucocorticoid activity
• It is useful to detect excess adrenal activity since it is produced nearly exclusively by the adrenal glands
Specifications

Specifications

Powder
Hygroscopic
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only