missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ LB Medium, Dehydrated (Capsules)

Formulations for the E. coli growth media known as LB Medium typically contain 10g of tryptone and 5g of yeast extract per liter, and LB Medium contains 10g NaCl per liter

Brand:  MP Biomedicals™ 113002011

69.03 EUR valid until 2024-12-31
Use promo code "21946" to get your promotional price.


 View more versions of this product

Product Code. 11373219

  • 72.70 € / 227g
Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Molecular Biology Certified™
Dehydrated
Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L
227 g
15°C to 30°C
Microbiology Media
For growth of Escherichia coli, plasmid and other bacterial cultures in liquid medium
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.