Learn More
LC3 Antibody Pack, Novus Biologicals™
Antibody Pack
Brand:  Novus Biologicals NB910-40435-1Pack
Product Code. 18598273
  • 647.00 € / 1 set
Explore more special offers
Description
Description
LC3 Antibody Packantibody specifically detects LC3 in Human, Mouse, Rat, Porcine, Alligator, Amphibian, Avian, Bacteria, Bovine, Canine, Chicken, Chinese Hamster, Fish, Guinea Pig, Hamster, Invertebrate, Plant, Primate, Monkey, Rabbit, Golden Syrian Hamster, Zebrafish samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Proximity Ligation Assay, SDS-Page, Southern Blot, Immunohistochemistry (Whole-Mount), ChIP assay (ChIP), Gene Knock-Out, KnockDown
Specifications
Specifications
LC3
Unconjugated
For NB100-2331: A synthetic peptide made to an internal portion of the human LC3 protein sequence (between residues 25-121) [Uniprot: Q9H492]., For NB100-2220: Polyclonal LC3B Antibody was made to a synthetic peptide made to an N-terminal portion of the human LC3B protein sequence (between residues 1-100) [UniProt# Q9GZQ8].
RUO
81631
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry
ATG8F, LC3B, MAP1A/1BLC3, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta
1 Pack
Autophagy, Cancer, Cardiovascular Biology, mTOR Pathway
Human, Mouse, Rat, Porcine, Alligator
Purified