Learn More
LC3 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-14600-100UL
Product Code. 17492493
  • 450.00 € / 100µL
Explore more special offers
Description
Description
The LC3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human LC3. This antibody recognizes human, mouse, rat antigen. The LC3 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™488.
Specifications
Specifications
LC3
Polyclonal
Rabbit
Primary
Human, Rat, Mouse
Flow Cytometry (Intracellular Staining)
CoraLite 488
100 μL
81631