missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14600-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody can cross react with MAP1LC3A, MAP1LC3B, and MAP1LC3C.
LC3B (Autophagy Marker Light Chain 3B) in humans, is encoded by the gene MAP1LC3B (Microtubule-associated proteins 1A/1B light chain 3B). The product of this gene is associated with microtubule assembly and important is in neurogenesis. Recent studies indicates that LC3B plays a vital role in autophagy, a process that involves the bulk degradation of cytoplasmic component. Three human LC3 isoforms undergo post-translational modifications during autophagy.Specifications
LC3 | |
Polyclonal | |
Unconjugated | |
MAP1LC3A | |
LC3, LC3B, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, MAP1A/MAP1B light chain 3 B, MAP1ALC3, MAP1LC3B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362245, 64862, 66734, 67443, 81631, 84557 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Flow Cytometry | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q62625, Q6XVN8, Q91VR7, Q9CQV6, Q9GZQ8, Q9H492 | |
Map1lc3a, Map1lc3b | |
LC3 Fusion Protein Ag6144 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction