Learn More
Testo™ LCD Display Thermo-Hygrometer
Use this thermohygrometer for a variety of applications. Testo™ 608-H1 Thermohygrometers provide a continuous display of current temperature and humidity values.
Brand:  Testo™ 0560 6081
Product Code. 10316802
  • 318.00 € / Each
Explore more special offers
Description
Description
  • Continuous measurement and display of temperature and relative humidity
  • Calculation and display of the dew point
  • Large, clear display
  • Capacitive humidity sensor with long-term stability for many years of measurement
Specifications
Specifications
Thermohygrometer
0°C to 50°C
168 g
For measure and display the temperature and relative humidity continuously
0.001
LCD
9 V