missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDLR Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10785-1-AP-150UL
This item is not returnable.
View return policy
Description
The low density lipoprotein receptor (LDL-R) gene family consists of cell surface proteins involved in receptor-mediated endocytosis of specific ligands. Low density lipoprotein (LDL) is normally bound at the cell membrane and taken into the cell ending up in lysosomes where the protein is degraded and the cholesterol is made available for repression of microsomal enzyme 3-hydroxy-3-methylglutaryl coenzyme A (HMG CoA) reductase, the rate-limiting step in cholesterol synthesis. At the same time, a reciprocal stimulation of cholesterol ester synthesis takes place. Mutations in the LDL-R gene cause the autosomal domit disorder, familial hypercholesterolemia. Along with SCARB1, CLDN1, and the tetraspanin superfamily member CD81, LDL-R has been reported to be an entry factor for the Hepatitis C virus. At least three isoforms of LDL-R are known to exist.Specifications
LDLR | |
Polyclonal | |
Unconjugated | |
LDLR | |
FH, FHC, LDL receptor, LDLR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16835, 3949 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P01130, P35951 | |
Ldlr | |
LDLR Fusion Protein Ag1236 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |