missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ LDPE Dropping Pipette

Low density polyethylene (LDPE) construction. Used for sampling and decanting infectious or toxic liquids. Graduated with bellows.

Brand:  BRAND™ 125420

29.60 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10604161

  • 44.20 € / Pack of 100
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • LDPE construction
  • Graduated
  • Includes bellows
  • For handling toxic or infections samples
  • Supplied in packs of 100
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LDPE
Yes
5 mL
Dropping Pipette
For Sampling and Decanting Infectious or Toxic Liquids
100/Pk.
194 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ LDPE Dropping Pipette > Capacity: 5mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.