missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Round Bucket with Stainless Steel Handle

Round Bucket with Stainless Steel Handle Food bucket and anti-wear skirt for increased product durability. Highly durable.

Brand:  Gilac™ G615128

 View more versions of this product

Product Code. 11742525

  • 24.50 € / Each
Estimated Shipment: 22-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Food bucket
  • Highly durable
  • Anti-wear skirt for increased product durability
  • Made from low density polyethylene making it highly resistant
  • Stainless steel handle
  • Reinforced Bottom
  • External dimensions include handles
Specifications

Specifications

Industrial Bucket
Yes
3.4 gal.
12 in.
Steel
310 mm
345 mm
-15°C to +60°C
305 mm
13 L
LDPE
Blue
305 mm
12.2 in.
13.6 in.
5°F to 140°F
12 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers