missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micronova™ LDPE/Nylon Blend Liner for Buckets
Designed for cleanrooms and controlled environments
Brand: Micronova™ BKL-2424IR
This item is not returnable.
View return policy
Description
- Irradiated
- 75% LDPE/25% cast nylon
- 10-6 sterility assurance level
- For cleanrooms and controlled environments
Certifications
ISO 9001:2008
Compliance
AAMI Guidelines for sterilization
Specifications
Blend Liner | |
LDPE, Nylon | |
Dimensions: 60.9 x 60.9 cm (24 x 24 in.) | |
Cleanrooms and Controlled Environments |