Learn More
Kautex™ LDPE Screw Closure
Feature LDPE light weight, tough construction. Kautex™ LDPE Screw Closures are available in a wide variety of sizes for Kautex™ narrow and wide neck bottles.
Brand: Kautex™ 2000090609
This item is not returnable.
View return policy
Description
- Made from low density polyethylene
- Translucent
- Available for a wide variety of sizes
Specifications
Natural | |
LDPE | |
500/750/1000 mL Wide Mouth Bottle | |
50 mm |