missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ LDPE Wash Bottles with Venting Valve

Product Code. 10489261
Click to view available options
Capacity:
250 mL
500 mL
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
This item is not returnable. View return policy
This item is not returnable. View return policy

High-quality wash bottles with integrated wash stream and sealing cone closure.

  • For distilled water and organic solvents only
  • Large opening for convenient filling
  • Label with designation of contents and general safety statements in English (was CAS numbers and NFPA codes) and German (was CAS numbers)
  • Manufactured from flexible PE-LD, limited to temperatures below 80°C

TRUSTED_SUSTAINABILITY

Specifications

Capacity 500 mL
Capacity (Metric) 500 mL
Material LDPE
Closure Color Red
Closure Included Yes
Mouth Wide
Product Type Wash Bottle
Chemical Label Acetone
Closure Type Stopper
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.