missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ LDPE Wash Bottles with Venting Valve
Click to view available options
Capacity:
250 mL
500 mL
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Description
- For distilled water and organic solvents only
- Large opening for convenient filling
- Label with designation of contents and general safety statements in English (was CAS numbers and NFPA codes) and German (was CAS numbers)
- Manufactured from flexible PE-LD, limited to temperatures below 80°C
Specifications
Specifications
Capacity | 500 mL |
Capacity (Metric) | 500 mL |
Material | LDPE |
Closure Color | Green |
Closure Included | Yes |
Mouth | Wide |
Product Type | Wash Bottle |
Chemical Label | Distilled Water |
Closure Type | Stopper |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction