missing translation for 'onlineSavingsMsg'
Learn More
LE 135, Tocris Bioscience™ Product Code.: 18005972

LE 135, Tocris Bioscience™

Product Code. 18005972
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18005972

Brand: Tocris 2021/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective RARβ antagonist

Retinoic acid antagonist; displays moderate selectivity for RARβ over RARα (Ki values are 0.22 and 1.4 μM respectively). Highly selective over RARγ and RXRα. Inhibits human HL-60 leukemia cell differentiation induced by Am80 (IC50 = 150 nM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155877-83-1
Molecular Formula C29H30N2O2
InChI Key YZZAIQOVMHVWBS-UHFFFAOYSA-N
PubChem CID 10410894

Specifications

Quantity 10 mg
Synonym chembl37708, 4-5,7,7,10,10-pentamethyl-8,9-dihydronaphtho 2,3-b 1, 4 benzodiazepin-13-yl benzoic acid, 4-5,7,7,10,10-pentamethyl-7,8,9,10-tetrahydro-5h-benzo e naphtho 2,3-b 1,4 diazepin-13-yl benzoic acid, d0x5wp, 4-7,8,9,10-tetrahydro-5,7,7,10,10-pentamethyl-5h-benzo e naphtho 2,3-b 1,4 diazepin-13-yl benzoic acid, 4-5,7,7,10,10-pentamethyl-7,8,9,10-tetrahydro-5 h-5,12-diaza-benzo 4,5 cyclohepta 1,2-b naphthalen-13-yl-benzoic acid, 4-5,7,7,10,10-pentamethyl-7,8,9,10-tetrahydro-5 h-5,12-diaza-benzo 4,5 cyclohepta 1,2-b naphthalen-13-yl-benzoic acid
SMILES CN1C2=CC=CC=C2C(=NC2=C1C=C1C(=C2)C(C)(C)CCC1(C)C)C1=CC=C(C=C1)C(O)=O
IUPAC Name 4-{2,14,14,17,17-pentamethyl-2,10-diazatetracyclo[9.8.0.0³,⁸.0¹³,¹⁸]nonadeca-1(11),3,5,7,9,12,18-heptaen-9-yl}benzoic acid
Molecular Weight (g/mol) 438.57
Formula Weight 438.56
Percent Purity >97%
Chemical Name or Material LE 135
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
LE 135, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.