missing translation for 'onlineSavingsMsg'
Learn More

Lead antimonide, 99+% (metals basis), Thermo Scientific™

87.90 € - 372.00 €

Chemical Identifiers

CAS 12266-38-5
Molecular Formula PbSb
Molecular Weight (g/mol) 328.96
MDL Number MFCD00151190
InChI Key QQHJESKHUUVSIC-UHFFFAOYSA-N
Synonym lead monoantimonide, lead antimonide, lead-antimonide, stibanylidyneplumbyl, antimony, compd. with lead 1:1, antimony, compound with lead 1:1
PubChem CID 72720464
IUPAC Name stibanylidynelead
SMILES [Sb]#[Pb]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11383438
View Documents
Thermo Scientific Alfa Aesar
036350.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11373438
View Documents
Thermo Scientific Alfa Aesar
036350.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

12266-38-5
328.96
QQHJESKHUUVSIC-UHFFFAOYSA-N
72720464
[Sb]#[Pb]
PbSb
MFCD00151190
lead monoantimonide, lead antimonide, lead-antimonide, stibanylidyneplumbyl, antimony, compd. with lead 1:1, antimony, compound with lead 1:1
stibanylidynelead

Specifications

12266-38-5
PbSb
UN1549
QQHJESKHUUVSIC-UHFFFAOYSA-N
stibanylidynelead
72720464
≥99%
6.72 g/mL
Lead antimonide
Lump
MFCD00151190
lead monoantimonide, lead antimonide, lead-antimonide, stibanylidyneplumbyl, antimony, compd. with lead 1:1, antimony, compound with lead 1:1
[Sb]#[Pb]
328.96
328.94
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c

H302+H332-H350-H360Df-H373

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.

missing translation for 'einecsNumber' : 235-536-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Lead antimonide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.