missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead foil, 1.6mm (0.063in) thick, 30x30cm, 99+% (metals basis), Thermo Scientific Chemicals
Form: Approx.1630g/30 x 30cm
198.00 € - 806.00 €
Chemical Identifiers
CAS | 7439-92-1 |
---|---|
Molecular Formula | Pb |
Molecular Weight (g/mol) | 207.20 |
MDL Number | MFCD00134050 |
InChI Key | WABPQHHGFIMREM-UHFFFAOYSA-N |
Synonym | plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant |
PubChem CID | 5352425 |
ChEBI | CHEBI:27889 |
IUPAC Name | lead |
SMILES | [Pb] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15435905
|
Thermo Scientific Alfa Aesar
000708.KF |
1 Pc. |
198.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15445905
|
Thermo Scientific Alfa Aesar
000708.KJ |
5 Pc. |
806.00 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-92-1 | |
207.20 | |
WABPQHHGFIMREM-UHFFFAOYSA-N | |
5352425 | |
lead |
Pb | |
MFCD00134050 | |
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant | |
CHEBI:27889 | |
[Pb] |
Spécification
Gray | |
Gray foil | |
Pb | |
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant | |
[Pb] | |
207.20 | |
CHEBI:27889 | |
Foil |
7439-92-1 | |
>99% (Metals basis) | |
MFCD00134050 | |
WABPQHHGFIMREM-UHFFFAOYSA-N | |
lead | |
5352425 | |
Odorless | |
Lead |
Sécurité et traitement
P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c
H350-H360FD-H362-H372
missing translation for 'einecsNumber' : 231-100-4
missing translation for 'rtecsNumber' : OF7525000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Identifiant produit
Mention d'avertissement
Catégorie de danger
Mention de danger
Conseils de prudence
- Lead foil
Mention d'avertissement
- Danger
Catégorie de danger
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Mention de danger
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Conseils de prudence
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only