Learn More
Lead(II) acetate basic, ACS reagent, Thermo Scientific Chemicals
58.69 € - 229.14 €
Chemical Identifiers
CAS | 51404-69-4 |
---|---|
Molecular Formula | C4H10O8Pb3 |
Molecular Weight (g/mol) | 807.69 |
InChI Key | XAOPZCQFBUCRKR-UHFFFAOYSA-L |
PubChem CID | 75087083 |
IUPAC Name | diacetyloxylead;lead;dihydrate |
SMILES | CC(=O)O[Pb]OC(=O)C.O.O.[Pb] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15254288
|
Thermo Scientific Acros
453220010 |
1 kg |
58.69 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
15264288
|
Thermo Scientific Acros
453220050 |
5 kg |
229.14 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
51404-69-4 | |
807.69 | |
75087083 | |
CC(=O)O[Pb]OC(=O)C.O.O.[Pb] |
C4H10O8Pb3 | |
XAOPZCQFBUCRKR-UHFFFAOYSA-L | |
diacetyloxylead;lead;dihydrate |
Specifications
51404-69-4 | |
XAOPZCQFBUCRKR-UHFFFAOYSA-L | |
diacetyloxylead;lead;dihydrate | |
75087083 | |
≥33.0% (basic Pb as PbO) | |
1.5% max. (105°C, 2 h) |
C4H10O8Pb3 | |
CC(=O)O[Pb]OC(=O)C.O.O.[Pb] | |
807.69 | |
807.69 | |
ACS Reagent | |
Lead(II) acetate basic |
Safety and Handling
GHS H Statement Harmful if swallowed. Harmful if inhaled. May damage the unborn child. Suspected of damaging fertility. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life with long lasting effects.
GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Obtain special instructions before use. Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 257-175-3
- Lead(II) acetate basic
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only