missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) acetate trihydrate, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals

Chemical Identifiers

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10035683
View Documents Promotion Details
Thermo Scientific Acros
317230050
5 g Glass bottle N/A
N/A
10606881
View Documents Promotion Details
Thermo Scientific Acros
317230250
25 g Glass bottle N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Chemical Identifiers

6080-56-4
379.30
MCEUZMYFCCOOQO-UHFFFAOYSA-L
22456
O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
C4H12O7Pb
MFCD00150023
lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
CHEBI:33112

Specifications

75.0°C
White
99.999%
(CH3CO2)2Pb·3H2O
01,532; 02,233; 04,276
lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
MCEUZMYFCCOOQO-UHFFFAOYSA-L
λ²-lead(2+) diacetate trihydrate
22456
379.33
Trace Metal Basis
6080-56-4
Crystalline Powder and/or Crystals
C4H12O7Pb
MFCD00150023
15, 5452
Solubility in water: 625g/L. Other solubilities: 1g/30mL alcohol, freely soluble in glycerol
O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
379.30
CHEBI:33112
99.999%
Lead(II) acetate trihydrate, 99.999%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause damage to organs through prolonged or repeated exposure if swallowed.
May damage the unborn child.
Suspected of damaging fertility.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHA

GHS Signal Word: Danger

missing translation for 'rtecsNumber' : OF8050000

Product Identifier
  • Lead(II) acetate trihydrate

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H318-Causes serious eye damage.
  • H350-May cause cancer.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only