missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) acetate trihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Chemical Identifiers

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10506802
View Documents Promotion Details
Thermo Scientific Acros
423840250
25 g Plastic bottle N/A
N/A
10639104
View Documents Promotion Details
Thermo Scientific Acros
423841000
100 g Plastic bottle N/A
N/A
10464321
View Documents Promotion Details
Thermo Scientific Acros
423845000
500 g Plastic bottle N/A
N/A
10629104
View Documents Promotion Details
Thermo Scientific Acros
423840025
2.5 kg Plastic bottle N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6080-56-4
379.30
MCEUZMYFCCOOQO-UHFFFAOYSA-L
22456
O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
C4H12O7Pb
MFCD00150023
lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
CHEBI:33112

Specifications

75.0°C
White
99.0 to 103% (Complexometry)
(CH3CO2)2Pb·3H2O
01,532; 02,233; 04,276
lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
MCEUZMYFCCOOQO-UHFFFAOYSA-L
λ²-lead(2+) diacetate trihydrate
22456
379.33
ACS Reagent
13.1
6080-56-4
Crystalline Powder and/or Crystals
C4H12O7Pb
MFCD00150023
15, 5452
Solubility in water: 625g/L. Other solubilities: 1g/30mL alcohol, freely soluble in glycerol
O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
379.30
CHEBI:33112
≥99%
Decomposes information: >100°C
Lead(II) acetate trihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause damage to organs through prolonged or repeated exposure if swallowed.
May damage the unborn child.
Suspected of damaging fertility.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHA

GHS Signal Word: Danger

missing translation for 'rtecsNumber' : OF8050000

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H318-Causes serious eye damage.
  • H350-May cause cancer.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only