missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) acetate trihydrate, 99%, Thermo Scientific Chemicals

Product Code. 11409293
Click to view available options
Quantity:
250 g
1000 g

Lead(II) acetate trihydrate is used as a mordant in textile printing and dyeing, drier in paints, varnishes and as a water repellant. It is also used in cosmetics and to prepare other lead compounds. It serves as an ingredient in progressive types of hair coloring dyes. Further, it is used in the detection of poisonous gas hydrogen sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lead(II) acetate trihydrate is used as a mordant in textile printing and dyeing, drier in paints, varnishes and as a water repellant. It is also used in cosmetics and to prepare other lead compounds. It serves as an ingredient in progressive types of hair coloring dyes. Further, it is used in the detection of poisonous gas hydrogen sulfide.

Solubility
Freely soluble in water, alcohol and glycerol.

Notes
Decomposes above 200°Air and light sensitive. Incompatible with strong acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
IUPAC Name lead(2+);diacetate;trihydrate
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Especificaciones

Melting Point 75°C
Density 2.55
Quantity 250 g
UN Number UN1616
Beilstein 3730298
Merck Index 14,5397
Solubility Information Freely soluble in water,alcohol and glycerol.
Formula Weight 379.33 (325.28 Anhydrous)
Percent Purity 99%
Chemical Name or Material Lead (II) acetate trihydrate
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Lead(II) acetate trihydrate
Palabra de advertencia
  • Danger
Clasificación
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Indicaciones de peligro
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Otros peligros
  • MIXTURE LIST-Contains : Lead (II) acetate, trihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'