missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Acetate Trihydrate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10548810
Click to view available options
Quantity:
250 g
500 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
IUPAC Name lead(2+);diacetate;trihydrate
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 75°C
Color White
pH 5.5 to 6.5
Packaging Plastic powder jar
Quantity 500 g
Formula Weight 379.33g/mol
Physical Form Solid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'