missing translation for 'onlineSavingsMsg'
Learn More

Lead(Ii) Acetate Trihydrate, puriss. p.a. ACS Reagent, Honeywell Fluka™

Puriss. p.a., ACS Reagent, Reagent ISO, Reagent European Pharmacopoeia, 99.5-102.0%

46.93 € - 129.21 €

Chemical Identifiers

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrateShow MoreShow Less
PubChem CID 22456
ChEBI CHEBI:33112
IUPAC Name lead(2+);diacetate;trihydrate
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15623800
View Documents
Honeywell-Fluka
32307-100G
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15643800
View Documents
Honeywell-Fluka
32307-250G
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15633800
View Documents
Honeywell-Fluka
32307-1KG
1 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead(Ii) Acetate Trihydrate, puriss. p.a. ACS Reagent, Honeywell Fluka™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.