missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) bromide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Lead(II) bromide, 99.999%, Br2Pb, CAS Number-10031-22-8, dibromoplumbane, acmc-1bq2x, lead 2+ bromide, ccris 4220, lead dibromide, lead bromide pbbr2, lead ii bromide, pbbr2, leadbromide, lead bromide, 25g, (metals basis), 916 deg.C, White, 6.66 g/mL

75.78 € - 251.25 €

Chemical Identifiers

CAS 10031-22-8
Molecular Formula Br2Pb
Molecular Weight (g/mol) 367.00
MDL Number MFCD00011156
InChI Key ZASWJUOMEGBQCQ-UHFFFAOYSA-L
Synonym lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
PubChem CID 24831
IUPAC Name dibromolead
SMILES [Br-].[Br-].[Pb++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11386787
SDS View Documents
Thermo Scientific Alfa Aesar
010720.06
5 g
75.78 €
5g
Please to purchase this item. Need a web account? Register with us today!
11396787
SDS View Documents
Thermo Scientific Alfa Aesar
010720.14
25 g
251.25 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used in the field of antirust, pigment and photograph. The molten lead(II) bromide acts as an electrolyte. Lead(II) bromide provides a high concentration of lead(II) ions and bromide ions to carry the current during the electrolysis process. The rare-earth-doped alkali-lead bromide crystals (potassium lead bromide (or) KPB; rubidium lead bromide or RPB) emerge as promising new low-phonon-energy host materials for mid-IR applications and are useful for solid state lasers. Hybrid organic/lead halide perovskites are promising materials for solar cell fabrication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10031-22-8
367.00
ZASWJUOMEGBQCQ-UHFFFAOYSA-L
24831
[Br-].[Br-].[Pb++]
Br2Pb
MFCD00011156
lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
dibromolead

Specifications

916°C
10031-22-8
Powder
PbBr2
UN2291
lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
ZASWJUOMEGBQCQ-UHFFFAOYSA-L
dibromolead
24831
99.999%
Odorless
(metals basis)
373°C
White
Br2Pb
MFCD00011156
14,5403
Slightly soluble in water; Insoluble in alcohol; Soluble in ammonia,alkali,KBr and NaBr.
[Br-].[Br-].[Pb++]
367.00
367.01
Puratronic™
6.66 g/mL
Lead(II) bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P312-P314-P330-P501c

H302+H332-H360Df-H373

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.

missing translation for 'einecsNumber' : 233-084-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Lead(II) bromide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.