missing translation for 'onlineSavingsMsg'
Learn More

Lead(Ii) Carbonate Basic, Honeywell Fluka™

Product Code. 15691050
Click to view available options
Quantity:
1 kg
50 kg
6 x 1 kg

Puriss., white lead,≥77% Pb basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1319-46-6
MDL Number MFCD00078155

Specifications

Quantity 1 kg
Linear Formula (PbCO3)2·Pb(OH)2
UN Number 3077
Formula Weight 775.63g/mol
Percent Purity ≥77% (Pb basis)
Grade puriss.
Chemical Name or Material Lead(Ii) Carbonate Basic
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) carbonate basic
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Lead(II) carbonate basic
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'