missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) carbonate, White powder, Puratronic™, 99.999% (Metals basis), Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
Color | White |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.999% (Metals basis) |
IUPAC Name | lead(2+);carbonate |
Formula Weight | 267.2 |
Odor | Odorless |
Chemical Name or Material | Lead(II) carbonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'