Learn More
Lead(II) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
Used in the synthesis of lead titanate and barium lead titanate ceramics; employed in the production of infrared transmitting glass and ornamental glass (aurene glass); useful as an electrode in geophysical applications and in solar cells.
69.35 € - 775.86 €
Chemical Identifiers
CAS | 7758-95-4 |
---|---|
Molecular Formula | Cl2Pb |
Molecular Weight (g/mol) | 278.10 |
MDL Number | MFCD00011157 |
InChI Key | HWSZZLVAJGOAAY-UHFFFAOYSA-L |
Synonym | lead dichloride, lead ii chloride, lead chloride, plumbous chloride, lead 2+ chloride, lead chloride pbcl2, pbcl2, ccris 7565, lead ii chloride, ultra dry |
PubChem CID | 24459 |
SMILES | [Cl-].[Cl-].[Pb++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11340058
|
Thermo Scientific Alfa Aesar
042841.04 |
2 g |
69.35 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11320058
|
Thermo Scientific Alfa Aesar
042841.09 |
10 g |
183.92 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11330058
|
Thermo Scientific Alfa Aesar
042841.18 |
50 g |
775.86 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lead chloride is used in the synthesis of lead titanate and barium lead titanate ceramics; employed in the production of infrared transmitting glass and ornamental glass (aurene glass); useful as an electrode in geophysical applications and in solar cells. Analytical reagent, preparation of lead salts, as solder and flux. Pattinson's white lead, pigment in white paint, is the basic lead chloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7758-95-4 | |
278.10 | |
HWSZZLVAJGOAAY-UHFFFAOYSA-L | |
24459 |
Cl2Pb | |
MFCD00011157 | |
lead dichloride, lead ii chloride, lead chloride, plumbous chloride, lead 2+ chloride, lead chloride pbcl2, pbcl2, ccris 7565, lead ii chloride, ultra dry | |
[Cl-].[Cl-].[Pb++] |
Specifications
951°C | |
7758-95-4 | |
Powder | |
Cl2Pb | |
lead dichloride, lead ii chloride, lead chloride, plumbous chloride, lead 2+ chloride, lead chloride pbcl2, pbcl2, ccris 7565, lead ii chloride, ultra dry | |
HWSZZLVAJGOAAY-UHFFFAOYSA-L | |
λ²-lead(2+) dichloride | |
24459 | |
Odorless | |
5.85 g/cm3 at 20°C |
501°C | |
White | |
99.999% (Metal basis) | |
MFCD00011157 | |
Soluble in hot water,alkali hydroxides and ammonium chloride solution. Insoluble in cold water and alcohol. | |
[Cl-].[Cl-].[Pb++] | |
278.10 | |
278.1 | |
White powder | |
Lead(II) chloride, ultra dry |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H350-H360Df-H372
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III
missing translation for 'einecsNumber' : 231-845-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Lead(II) chloride, ultra dry
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.