missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) fluoride, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
Description
Lead(II) fluoride is used in fuses, glass coatings to reflect infrared rays and phosphors for television screens and in low melting glasses. Further, it serves as a catalyst for the preparation of picoline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 1290°C |
Melting Point | 855°C |
Physical Form | Powder |
Quantity | 100 g |
UN Number | UN2291 |
Merck Index | 14,5408 |
Solubility Information | Soluble in nitric acid. Slightly soluble in water. Insoluble in acetone and ammonia. |
Formula Weight | 245.2 |
Percent Purity | 99.997% |
Grade | Puratronic™ |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'