missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) fluoride, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Product Code. 15455965
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Lead(II) fluoride is used in fuses, glass coatings to reflect infrared rays and phosphors for television screens and in low melting glasses. Further, it serves as a catalyst for the preparation of picoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-46-2
Molecular Formula F2Pb
Molecular Weight (g/mol) 245.20
MDL Number MFCD00011162
InChI Key FPHIOHCCQGUGKU-UHFFFAOYSA-L
Synonym lead fluoride, lead ii fluoride, lead difluoride, lead fluoride pbf2, plumbous fluoride, lead +2 fluoride, difluoroplumbane, plomb fluorure french, pbf2, plomb fluorure
PubChem CID 24549
IUPAC Name difluorolead
SMILES [F-].[F-].[Pb++]

Specifications

Boiling Point 1290°C
Melting Point 855°C
Physical Form Powder
Quantity 25 g
UN Number UN2291
Merck Index 14,5408
Solubility Information Soluble in nitric acid. Slightly soluble in water. Insoluble in acetone and ammonia.
Formula Weight 245.2
Percent Purity 99.997%
Grade Puratronic™
Odor Odorless
Density 8.445 g/mL
Assay (metals basis)
Chemical Name or Material Lead(II) fluoride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) fluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.