missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) hexafluorosilicate dihydrate, Thermo Scientific™

25.50 € - 84.50 €

Chemical Identifiers

CAS 1310-03-8
Molecular Formula F6H4O20Pb9Si6
Molecular Weight (g/mol) 2471.312
MDL Number MFCD00049509
InChI Key UCXBPZOKVHNQLY-UHFFFAOYSA-N
Synonym acmc-20aknf, fluoro trioxido silane;lead 2+ ;dihydrate, silicate 2-,hexafluoro-, lead 2+ 1:1 , dihydrate 9ci
PubChem CID 57347846
IUPAC Name fluoro(trioxido)silane;lead(2+);dihydrate
SMILES O.O.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11378108
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039347.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11368108
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039347.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1310-03-8
2471.312
UCXBPZOKVHNQLY-UHFFFAOYSA-N
57347846
O.O.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2]
F6H4O20Pb9Si6
MFCD00049509
acmc-20aknf, fluoro trioxido silane;lead 2+ ;dihydrate, silicate 2-,hexafluoro-, lead 2+ 1:1 , dihydrate 9ci
fluoro(trioxido)silane;lead(2+);dihydrate

Specifications

1310-03-8
F6H4O20Pb9Si6
MFCD00049509
acmc-20aknf, fluoro trioxido silane;lead 2+ ;dihydrate, silicate 2-,hexafluoro-, lead 2+ 1:1 , dihydrate 9ci
O.O.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2]
2471.312
385.26 (349.26 Anhydrous)
Lead(II) hexafluorosilicate dihydrate
Powder
PbSiF6·2H2O
UN2291
UCXBPZOKVHNQLY-UHFFFAOYSA-N
fluoro(trioxido)silane;lead(2+);dihydrate
57347846
Odorless
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c

H302+H332-H350-H360Df-H373

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.

missing translation for 'einecsNumber' : 247-278-1

missing translation for 'rtecsNumber' : VV8450000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Lead(II) hexafluorosilicate dihydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications