missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) iodide, 99%, Thermo Scientific Chemicals

Product Code. 10123740
50 g, Glass bottle
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-63-0
Molecular Formula I2Pb
Molecular Weight (g/mol) 461.00
MDL Number MFCD00011163
InChI Key RQQRAHKHDFPBMC-UHFFFAOYSA-L
Synonym lead ii iodide, lead diiodide, plumbous iodide, lead iodide pbi2, pbi2, plumbum jodatum, blei ii-iodid, hsdb 636, diiodo-, e2-plumbane
PubChem CID 24931
IUPAC Name λ²-lead(2+) diiodide
SMILES [I-].[I-].[Pb++]

Specifications

Melting Point 402°C
Color Yellow to Orange
Physical Form Powder
Quantity 50 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula PbI2
Merck Index 15, 5466
Formula Weight 461
Percent Purity 99%
Loss on Drying 0.1% max.
Packaging Glass bottle
Specific Gravity 6.16
Density 6.1600g/mL
Chemical Name or Material Lead(II) iodide, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467718 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number198862500,198860500
Lot NumberA0464943 Certificate TypeCertificate of Analysis Date08/08/2024 Catalog Number198862500,198860500
Lot NumberA0459426 Certificate TypeCertificate of Analysis Date23/01/2024 Catalog Number198862500,198860500
Lot NumberA0452975 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number198862500,198860500
Lot NumberA0439681 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number198862500,198860500
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) iodide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead(II) iodide, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.