Learn More
Lead(II,IV) oxide, red, 98%, Thermo Scientific Chemicals
23.62 € - 156.78 €
Chemical Identifiers
CAS | 1314-41-6 |
---|---|
Molecular Formula | O4Pb3 |
Molecular Weight (g/mol) | 685.57 |
MDL Number | MFCD00078491 |
InChI Key | XMFOQHDPRMAJNU-UHFFFAOYSA-N |
Synonym | lead tetroxide, orange lead, lead oxide, red, lead ii,iv oxide, po4b3, lead oxide, red, lr, lead oxide, red powder, 1-2 mum, lead oxide, red, saj first grade, lead oxide, powder trace metals basis |
PubChem CID | 16685188 |
IUPAC Name | 1,3,5,7-tetraoxa-2$l^{2},4,6$l^{2}-triplumbaspiro[3.3]heptane |
SMILES | O1[Pb]O[Pb]12O[Pb]O2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10781301
|
Thermo Scientific Acros
|
5 g | Glass bottle |
N/A
|
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
10626052
|
Thermo Scientific Acros
221111000 |
100 g | Glass bottle |
23.62 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10430881
|
Thermo Scientific Acros
221115000 |
500 g | Glass bottle |
34.57 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15159243
|
Thermo Scientific Acros
221110051 |
5 kg | Glass bottle |
156.78 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
1314-41-6 | |
685.57 | |
XMFOQHDPRMAJNU-UHFFFAOYSA-N | |
16685188 | |
O1[Pb]O[Pb]12O[Pb]O2 |
O4Pb3 | |
MFCD00078491 | |
lead tetroxide, orange lead, lead oxide, red, lead ii,iv oxide, po4b3, lead oxide, red, lr, lead oxide, red powder, 1-2 mum, lead oxide, red, saj first grade, lead oxide, powder trace metals basis | |
1,3,5,7-tetraoxa-2$l^{2},4,6$l^{2}-triplumbaspiro[3.3]heptane |
Specifications
500.0°C | |
Orange | |
97.5% min. (PB3O4) | |
2PbO·PbO2 | |
15,548 | |
Solubility in water: insoluble. Other solubilities: partly soluble in acids, insoluble in alcohol | |
O1[Pb]O[Pb]12O[Pb]O2 | |
685.57 | |
685.57 | |
Lead(II, IV) oxide, red, 98% |
1314-41-6 | |
Powder | |
O4Pb3 | |
MFCD00078491 | |
lead tetroxide, orange lead, lead oxide, red, lead ii,iv oxide, po4b3, lead oxide, red, lr, lead oxide, red powder, 1-2 mum, lead oxide, red, saj first grade, lead oxide, powder trace metals basis | |
XMFOQHDPRMAJNU-UHFFFAOYSA-N | |
1,3,5,7-tetraoxa-2$l^{2},4,6$l^{2}-triplumbaspiro[3.3]heptane | |
16685188 | |
98% |
Safety and Handling
GHS H Statement
Causes damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Harmful if swallowed.
May damage the unborn child.
Suspected of damaging fertility.
Suspected of causing cancer.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and k
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 215-235-6
missing translation for 'rtecsNumber' : OG5425000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.